missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-nitrobenzoic acid, 97%

Product Code. p-7052449
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15426617

Brand: Thermo Scientific Alfa Aesar H33757.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116465-48-6 | C7H3F2NO4 | 203.101 g/mol

2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116465-48-6
Molecular Formula C7H3F2NO4
Molecular Weight (g/mol) 203.101
MDL Number MFCD03425705
InChI Key GPTNSBLYGCZJQV-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-nitrobenzoic acid, 2,5-difluoro-4-nitrobenzenecarboxylic acid, 2,5-difluoro-4-nitrobenzic acid, 2,5-difluoro-4-nitrobenzoicacid, 2,5-difluoro-4-nitro-benzoic acid, benzoic acid, 2,5-difluoro-4-nitro, 2,5-difluoro-4-nitrobenzenecarboxylicacid, pubchem9789, acmc-2099sr, ksc495m6l
PubChem CID 246375
SMILES C1=C(C(=CC(=C1F)[N+](=O)[O-])F)C(=O)O

Specifications

Melting Point 145°C to 147°C
Color Green to White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 203.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Difluoro-4-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.