missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-hydroxybenzaldehyde, 99%

Product Code. 15464127
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464127 5 g 5g
15474127 250 mg 250mg
11345145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15464127 Supplier Thermo Scientific Alfa Aesar Supplier No. H31528.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 918523-99-6 | C7H4F2O2 | 158.104 g/mol

2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 918523-99-6
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD04112521
InChI Key FFPHRYLGIPAZBL-UHFFFAOYSA-N
Synonym 4-hydroxy-2,5-difluorobenzaldehyde, 2,5-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,5-difluoro-4-hydroxy, ksc496a0b
PubChem CID 21966462
IUPAC Name 2,5-difluoro-4-hydroxybenzaldehyde
SMILES C1=C(C(=CC(=C1F)O)F)C=O

Specifications

Melting Point 145°C to 148°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 158.11
Percent Purity 99%
Chemical Name or Material 2,5-Difluoro-4-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.