missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorotoluene, 98%, Thermo Scientific™

Product Code. 10696292 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10696292 5 g Glass bottle 5g
10050820 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10696292 Supplier Thermo Scientific Acros Supplier No. 113780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19398-61-9
MDL Number MFCD00000609
InChI Key KFAKZJUYBOYVKA-UHFFFAOYSA-N
Synonym 2,5-dichlorotoluene, benzene, 1,4-dichloro-2-methyl, toluene, 2,5-dichloro, 1,4-dichloro-2-methyl-benzene, toluene,5-dichloro, pubchem3700, acmc-209exk, ksc490m7d, benzene,4-dichloro-2-methyl
PubChem CID 29572
IUPAC Name 1,4-dichloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6Cl2
Linear Formula CH3C6H3Cl2
Refractive Index 1.5460 to 1.5480 (20°C, 589nm)
Quantity 5 g
Beilstein 05, 296
Molecular Weight (g/mol) 161.025
Formula Weight 161.03
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2, 5-Dichlorotoluene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.