missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-carboxylic acid, 98+%, Thermo Scientific™

Product Code. 11339645
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339645 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11339645 Supplier Thermo Scientific Alfa Aesar Supplier No. L19101.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 36157-41-2
Melting Point 147°C to 149°C
Molecular Formula C5H2Cl2O2S
MDL Number MFCD00082908
Quantity 2 g
Beilstein 127865
Synonym 2,5-dichloro-3-thiophenecarboxylic acid, 2,5-dichlorothiophene-3-carboxylicacid, 3-thiophenecarboxylic acid, 2,5-dichloro, acmc-1ajgc, ksc222q1l, 2,5-dichloro-thiophene-3-carboxylic acid, 3-thiophenecarboxylicacid, 2,5-dichloro, 2,5-dichlorothiophene-3-carboxylic acid
InChI Key FBUUZRITKBLZJX-UHFFFAOYSA-N
SMILES C1=C(SC(=C1C(=O)O)Cl)Cl
IUPAC Name 2,5-dichlorothiophene-3-carboxylic acid
Molecular Weight (g/mol) 197.029
PubChem CID 225438
Formula Weight 197.04
Percent Purity ≥98%
Chemical Name or Material 2,5-Dichlorothiophene-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.