missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine, 98%

Product Code. 11494070
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494070 25 g 25g
11442797 50 g 50g
11481960 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494070 Supplier Thermo Scientific Alfa Aesar Supplier No. A15442.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16110-09-1 | C5H3Cl2N | 147.99 g/mol

2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16110-09-1
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006239
InChI Key GCTFDMFLLBCLPF-UHFFFAOYSA-N
Synonym pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
PubChem CID 27685
IUPAC Name 2,5-dichloropyridine
SMILES ClC1=CC=C(Cl)N=C1

Specifications

Melting Point 59°C to 62°C
Boiling Point 190°C to 191°C
Flash Point 112°C (233°F)
Odor Unpleasant
Quantity 25 g
UN Number UN2811
Beilstein 108886
Solubility Information Insoluble in water.
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2,5-Dichloropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.