missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine, 98%

Product Code. 10104400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104400 25 g Glass bottle 25g
10367220 5 g Glass bottle 5g
10301614 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10104400 Supplier Thermo Scientific Acros Supplier No. 186140250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16110-09-1 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16110-09-1
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006239
InChI Key GCTFDMFLLBCLPF-UHFFFAOYSA-N
Synonym pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
PubChem CID 27685
IUPAC Name 2,5-dichloropyridine
SMILES ClC1=CC=C(Cl)N=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color White to Yellow
Density 112g/mL
Boiling Point 190.0°C to 191.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Cl2
Quantity 25 g
Beilstein 20, I, 80
Solubility Information Solubility in water: insoluble
Formula Weight 147.99
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2, 5-Dichloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.