missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine-3-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10131042
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10131042 10 g 10g
10099491 1 g 1g
10742947 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10131042 Supplier Thermo Scientific Maybridge Supplier No. SPB06842EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 78686-87-0
Molecular Formula C6H2Cl3NO
Molecular Weight (g/mol) 210.438
MDL Number MFCD00210149
InChI Key VRJMAKCJTSZUQR-UHFFFAOYSA-N
Synonym 2,5-dichloronicotinoyl chloride, 2,5-dichloro-nicotinoyl chloride, 2,5-dichloro-3-pyridinecarbonyl chloride, 2,5-dichloronicotinic acid chloride, 2,5-dichloropyridine-3-carbonylchloride, 3-chlorocarbonyl-2,5-dichloropyridine, 2,5-dichloro-pyridine-3-carbonyl chloride, 3-pyridinecarbonylchloride, 2,5-dichloro, 3-pyridinecarbonyl chloride, 2,5-dichloro
PubChem CID 2799996
IUPAC Name 2,5-dichloropyridine-3-carbonyl chloride
SMILES C1=C(C(=NC=C1Cl)Cl)C(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 210.45
Percent Purity 97%
Chemical Name or Material 2, 5-Dichloropyridine-3-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.