missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridin-3-amine, ≥95%, Thermo Scientific™

Product Code. 10315542
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10315542

Brand: Thermo Scientific Maybridge SPB06843DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 78607-32-6
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C5H4Cl2N2
MDL Number MFCD00204157
Quantity 1 g
Synonym 3-amino-2,5-dichloropyridine, 2,5-dichloro-3-aminopyridine, 2,5-dichloro-pyridin-3-ylamine, 3-pyridinamine, 2,5-dichloro, 2,5-dichloro-3-pyridylamine, 2,5-dichloropyridine-3-amine, pubchem5507, acmc-209per, ksc495k8j
InChI Key OLZPJUVEGSNIJL-UHFFFAOYSA-N
SMILES C1=C(C(=NC=C1Cl)Cl)N
IUPAC Name 2,5-dichloropyridin-3-amine
Molecular Weight (g/mol) 163.001
PubChem CID 2799999
Formula Weight 163.01
Percent Purity 95%
Chemical Name or Material 2, 5-Dichloropyridin-3-amine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.