missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenylhydrazine, 98%, Thermo Scientific™

Product Code. 11376966
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11376966

Brand: Thermo Scientific Alfa Aesar L06708.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-15-7
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.028
MDL Number MFCD00007576
InChI Key LZKWWERBNXLGLI-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl hydrazine, hydrazine, 2,5-dichlorophenyl, 2,5-dichloro-phenyl-hydrazine, 1-2,5-dichlorophenyl hydrazine, acmc-209hgk, wln: zmr bg eg, 2,5-dichloro-phenylhydrazine, hydrazine,5-dichlorophenyl, 0-15-00-00431 beilstein handbook reference
PubChem CID 9366
IUPAC Name (2,5-dichlorophenyl)hydrazine
SMILES C1=CC(=C(C=C1Cl)NN)Cl

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
Beilstein 743382
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorophenylhydrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.