missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenylboronic Acid, TRC

Product Code. 30388620
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388620 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388620 Supplier Toronto Research Chemicals Supplier No. TRCD43655025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dichlorophenylboronic Acid
CAS 135145-90-3
Molecular Formula C6H5BCl2O2
Quantity 25 g
Synonym B-(2,5-Dichlorophenyl)boronic Acid, 2,5-Dichlorobenzeneboronic Acid, (2,5-Dichlorophenyl)boronic Acid
InChI Formula InChI=1S/C6H5BCl2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,10-11H
SMILES OB(O)c1cc(Cl)ccc1Cl
IUPAC Name (2,5-dichlorophenyl)boronic acid
Molecular Weight (g/mol) 190.82
Formula Weight 189.976
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.