missing translation for 'onlineSavingsMsg'
Learn More

2,5-dichlorophenyl isothiocyanate, Thermo Scientific™

Product Code. 10515004
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3386-42-3
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
InChI Key JHTPBGFVWWSHDL-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl isothiocyanate, 2,5-dichlorophenylisothiocyanate, acmc-1cjox, 2,5-dichlorobenzenisothiocyanate, #, benzene,1,4-dichloro-2-isothiocyanato, benzene, 1,4-dichloro-2-isothiocyanato, 1,4-bis chloranyl-2-isothiocyanato-benzene
PubChem CID 137915
IUPAC Name 1,4-dichloro-2-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)N=C=S)Cl

Specifications

Assay Percent Range 97+%
Quantity 25 g
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2, 5-Dichlorophenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'