missing translation for 'onlineSavingsMsg'
Learn More

2,5-dichlorophenyl isothiocyanate, Thermo Scientific™

Product Code. 10026684
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10026684 5 g 5g
10557363 10 g 10g
10515004 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10026684 Supplier Thermo Scientific Maybridge Supplier No. SB01698DE
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3386-42-3
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
InChI Key JHTPBGFVWWSHDL-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl isothiocyanate, 2,5-dichlorophenylisothiocyanate, acmc-1cjox, 2,5-dichlorobenzenisothiocyanate, #, benzene,1,4-dichloro-2-isothiocyanato, benzene, 1,4-dichloro-2-isothiocyanato, 1,4-bis chloranyl-2-isothiocyanato-benzene
PubChem CID 137915
IUPAC Name 1,4-dichloro-2-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)N=C=S)Cl

Specifications

Assay Percent Range 97+%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2, 5-Dichlorophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.