missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenol, 98%

Product Code. 11406536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406536 25 g 25g
11416536 100 g 100g
11426536 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406536 Supplier Thermo Scientific Alfa Aesar Supplier No. B24787.14
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-78-8
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002174
InChI Key RANCECPPZPIPNO-UHFFFAOYSA-N
Synonym phenol, 2,5-dichloro, 2,5-dichloro-phenol, 2,5-dcp, unii-3b11g9akba, 3,6-dichlorophenol, ccris 5903, 3b11g9akba, dsstox_cid_5003, dsstox_rid_77621, dsstox_gsid_25003
PubChem CID 66
ChEBI CHEBI:27929
IUPAC Name 2,5-dichlorophenol
SMILES OC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 55°C to 58°C
Boiling Point 211°C
Odor Phenol-like
Quantity 25 g
UN Number UN2020
Beilstein 1907692
Formula Weight 163
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.