missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenol, 98%

Product Code. 10275371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275371 250 g Glass bottle 250g
10748732 50 g Glass bottle 50g
13498308 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10275371 Supplier Thermo Scientific Acros Supplier No. 113582500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-78-8
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002174
InChI Key RANCECPPZPIPNO-UHFFFAOYSA-N
Synonym phenol, 2,5-dichloro, 2,5-dichloro-phenol, 2,5-dcp, unii-3b11g9akba, 3,6-dichlorophenol, ccris 5903, 3b11g9akba, dsstox_cid_5003, dsstox_rid_77621, dsstox_gsid_25003
PubChem CID 66
ChEBI CHEBI:27929
IUPAC Name 2,5-dichlorophenol
SMILES OC1=CC(Cl)=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color White to Brown
Boiling Point 211.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 250 g
Beilstein 06,189
Solubility Information Solubility in water: 3g/L (20°C)
Formula Weight 163
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 2, 5-Dichlorophenol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dichlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.