missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloronitrobenzene, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 2,5-Dichloronitrobenzene |
| CAS | 89-61-2 |
| Molecular Formula | C6 H3 Cl2 N O2 |
| Quantity | 25 g |
| Synonym | Benzene, 1,4-dichloro-3-nitro- (1CI), 1,4-Dichloro-2-nitrobenzene, 1,4-Dichloro-3-nitrobenzene, 1-Nitro-2,5-dichlorobenzene, 2,5-Dichloro-1-nitrobenzene, 2,5-Dichloronitrobenzene, 2-Nitro-1,4-dichlorobenzene, NSC 406125, NSC 6294, Nitro-p-dichlorobenzene |
| InChI Formula | InChI=1S/C6H3Cl2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H |
| SMILES | [O-][N+](=O)c1cc(Cl)ccc1Cl |
| IUPAC Name | 1,4-dichloro-2-nitrobenzene |
| Molecular Weight (g/mol) | 192 |
| Formula Weight | 190.9541 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction