missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloronitrobenzene, Mikromol™

Product Code. 30052431
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052431

Brand: Mikromol MM1273.220025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,5-Dichloronitrobenzene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dichloronitrobenzene
CAS 89-61-2
Analyte or Component Names 2,5-Dichloronitrobenzene
Shipping Condition Room Temperature
Molecular Formula C6 H3 Cl2 N O2
Certifications/Compliance ISO 17025
Synonym Benzene, 1,4-dichloro-2-nitro-, Benzene, 1,4-dichloro-3-nitro- (1CI), 1,4-Dichloro-2-nitrobenzene, 1,4-Dichloro-3-nitrobenzene, 1-Nitro-2,5-dichlorobenzene, 2,5-Dichloro-1-nitrobenzene, 2,5-Dichloronitrobenzene, 2-Nitro-1,4-dichlorobenzene, NSC 406125, NSC 6294, Nitro-p-dichlorobenzene
InChI Formula InChI=1S/C6H3Cl2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H
SMILES [O-][N+](=O)c1cc(Cl)ccc1Cl
IUPAC Name 1,4-dichloro-2-nitrobenzene
Molecular Weight (g/mol) 192
Quantity 25 mg
Formula Weight 190.9541 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.