missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloronicotinic acid, 98%, Thermo Scientific™

Product Code. 15462439
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59782-85-3
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD01861979
InChI Key SXQSMLIMBNMUNB-UHFFFAOYSA-N
Synonym 2,5-dichloronicotinic acid, 2,5-dichloronicotinicacid, 3-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-3-pyridinecarboxylic acid, pubchem2601, acmc-209mfn, pyni16, 2,5-dichloro-nicotinic acid, 2,5-dichloronicotinic acid?, ksc269g2r
PubChem CID 2736085
IUPAC Name 2,5-dichloropyridine-3-carboxylic acid
SMILES C1=C(C(=NC=C1Cl)Cl)C(=O)O

Specifications

Melting Point 156°C to 158°C
Color White
Density 1.61 g/mL
Quantity 1 g
Beilstein 389586
Formula Weight 192
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,5-Dichloronicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'