missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzylamine, 97%

Product Code. 11318535
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318535 5 g 5g
11308535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318535 Supplier Thermo Scientific Alfa Aesar Supplier No. L11972.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10541-69-2 | C7H7Cl2N | 176.04 g/mol

2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Incompatible with strong oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10541-69-2
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00052391
InChI Key AKGJLIXNRPNPCH-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzylamine, 2,5-dichlorophenyl methanamine, benzenemethanamine, 2,5-dichloro, 2,5-dichloro-benzylamine, 1-2,5-dichlorophenyl methanamine, 2,5-dichlorophenyl methylamine, 2,5-dichlorobenzylam, acmc-1bxya, rarechem al bw 0384, 2,5-dichlorobenzyl amine
PubChem CID 457600
IUPAC Name (2,5-dichlorophenyl)methanamine
SMILES C1=CC(=C(C=C1Cl)CN)Cl

Specifications

Density 1.318
Boiling Point 74°C to 76°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.5775
Quantity 5 g
UN Number UN2735
Beilstein 2639302
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.