missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl Chloride, TRC

Product Code. 30377487
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377487 1 g
30369742 500 mg
30387500 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377487 Supplier Toronto Research Chemicals Supplier No. TRCD4384101G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2745-49-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
Synonym 1,4-Dichloro-2-(chloromethyl)benzene, (2,5-Dichlorophenyl)methyl Chloride, a,2,5-Trichlorotoluene
SMILES ClCc1cc(Cl)ccc1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5Cl3/c8-4-5-3-6(9)1-2-7(5)10/h1-3H,4H2
Formula Weight 193.9457
Chemical Name or Material 2,5-Dichlorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.