missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl chloride, 97%

Product Code. 11311937
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311937 5 g 5g
11321937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311937 Supplier Thermo Scientific Alfa Aesar Supplier No. L07506.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2745-49-5 | C7H5Cl3 | 195.467 g/mol

2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2745-49-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00035818
InChI Key OMZINLIPPVNUOG-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzyl chloride, 1,4-dichloro-2-chloromethyl benzene, 2,5-dichlorobenzylchloride, 1,4-dichloro-2-chloromethyl-benzene, unii-g46i9555fb, benzene, 1,4-dichloro-2-chloromethyl, acmc-20a58t, alpha,2,5-trichlorotoluene, unii-6q4qcu6n2r component
PubChem CID 75976
IUPAC Name 1,4-dichloro-2-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)CCl)Cl

Specifications

Boiling Point 121°C to 124°C (14 mmHg)
Odor Pungent
Refractive Index 1.5755
Quantity 5 g
UN Number UN3265
Beilstein 637802
Solubility Information Not miscible in water.
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.