missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 11312037
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34145-05-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004607
InChI Key LCEIGNVIDJNUGF-UHFFFAOYSA-N
PubChem CID 118604
IUPAC Name (2,5-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)CO)Cl

Specifications

Melting Point 76°C to 80°C
Quantity 5 g
Beilstein 3240770
Synonym 2,5-dichlorophenyl methanol, 2,5-dichlorobenzyl alcohol, benzenemethanol, 2,5-dichloro, 2,5-dichlorobenzylic alcohol, 2,5-dichloro-phenyl-methanol, 2,5-dichlorobenzylalcohol, acmc-1ad0a, 2,5-dichloro benzylalcohol, 2,5-dichloro-benzyl alcohol, benzenemethanol,2,5-dichloro
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,5-Dichlorobenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'