Learn More
2,5-Dichlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
£19.50 - £54.50
Chemical Identifiers
CAS | 320-50-3 |
---|---|
Molecular Formula | C7H3Cl2F3 |
Molecular Weight (g/mol) | 214.996 |
MDL Number | MFCD00000608 |
InChI Key | DYBYUWVMLBBEMA-UHFFFAOYSA-N |
Synonym | 2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene |
PubChem CID | 35360 |
IUPAC Name | 1,4-dichloro-2-(trifluoromethyl)benzene |
SMILES | C1=CC(=C(C=C1Cl)C(F)(F)F)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423704
|
Thermo Scientific Alfa Aesar
A11310.06 |
5 g |
£19.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433704
|
Thermo Scientific Alfa Aesar
A11310.14 |
25 g |
£54.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
320-50-3 | |
214.996 | |
DYBYUWVMLBBEMA-UHFFFAOYSA-N | |
35360 | |
C1=CC(=C(C=C1Cl)C(F)(F)F)Cl |
C7H3Cl2F3 | |
MFCD00000608 | |
2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene | |
1,4-dichloro-2-(trifluoromethyl)benzene |
Specifications
320-50-3 | |
76°C (168°F) | |
1.483 | |
UN1760 | |
2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene | |
C1=CC(=C(C=C1Cl)C(F)(F)F)Cl | |
214.996 | |
215 | |
2,5-Dichlorobenzotrifluoride |
1.483 | |
C7H3Cl2F3 | |
MFCD00000608 | |
1955442 | |
DYBYUWVMLBBEMA-UHFFFAOYSA-N | |
1,4-dichloro-2-(trifluoromethyl)benzene | |
35360 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 206-276-0
missing translation for 'rtecsNumber' : CZ5553755
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only