missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzenesulfonic acid dihydrate, 97%, Thermo Scientific™

Product Code. 11319919
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319919 100 g 100g
11309919 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319919 Supplier Thermo Scientific Alfa Aesar Supplier No. B25650.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38484-94-5
Molecular Formula C6H8Cl2O5S
Molecular Weight (g/mol) 263.085
MDL Number MFCD08271971
InChI Key VPJICTWHRNQYRG-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzenesulfonic acid dihydrate, acmc-209qt1, 2,5-dichlorobenzene-1-sulfonic acid-water 1/2
PubChem CID 21114734
IUPAC Name 2,5-dichlorobenzenesulfonic acid;dihydrate
SMILES C1=CC(=C(C=C1Cl)S(=O)(=O)O)Cl.O.O

Specifications

Melting Point 98°C to 102°C
Quantity 100 g
UN Number UN2585
Formula Weight 263.09 (227.07 Anhydrous)
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzenesulfonic acid dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.