missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzeneboronic acid, 98%

Product Code. 11426146
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426146 1 g 1g
11436146 5 g 5g
11446146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426146 Supplier Thermo Scientific Alfa Aesar Supplier No. B22984.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 135145-90-3 | C6H5BCl2O2 | 190.814 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135145-90-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.814
MDL Number MFCD01863182
InChI Key NNTFPBXQPOQRBT-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzeneboronic acid, 2,5-dichlorophenyl boronic acid, 2,5-dichlorophenylboronicacid, 2,5-dichlorophenyl boranediol, boronic acid, 2,5-dichlorophenyl, pubchem1811, acmc-209bxy, ksc174k3h, 2,5-dichloro-phenylboronic acid
PubChem CID 2773371
IUPAC Name (2,5-dichlorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)Cl)Cl)(O)O

Specifications

Melting Point ∼210°C to 216°C
Quantity 1 g
Beilstein 4801290
Formula Weight 190.82
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.