missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroaniline, TRC

Product Code. 30384005
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384005 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384005 Supplier Toronto Research Chemicals Supplier No. TRCD43171050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 95-82-9
Molecular Formula C6 H5 Cl2 N
Quantity 50 g
Synonym Benzenamine, 2,5-dichloro-, 2,5-DCA
InChI Formula InChI=1S/C6H5Cl2N/c7-4-1-2-5(8)6(9)3-4/h1-3H,9H2
SMILES Nc1cc(Cl)ccc1Cl
IUPAC Name 2,5-dichloroaniline
Molecular Weight (g/mol) 162.02
Formula Weight 160.9799
Chemical Name or Material 2,5-Dichloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.