missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dichloroacetophenone, 98%

Product Code. 11464824
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464824 100 g 100g
11424080 10 g 10g
11454824 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464824 Supplier Thermo Scientific Alfa Aesar Supplier No. A13770.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2476-37-1 | C8H6Cl2O | 189.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2476-37-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000607
InChI Key CYNFEPKQDJHIMV-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
PubChem CID 75587
IUPAC Name 1-(2,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 11°C to 13°C
Density 1.312
Boiling Point 118°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.561
Quantity 100 g
Beilstein 1865043
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',5'-Dichloroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.