missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dichloroacetophenone, 96%, Thermo Scientific™

Product Code. 15177412
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2476-37-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000607
InChI Key CYNFEPKQDJHIMV-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
PubChem CID 75587
IUPAC Name 1-(2,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Cl)Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Brown to Yellow
Density 1.3120g/mL
Boiling Point 118.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3COCH3
Refractive Index 1.5604 to 1.5624
Quantity 10 g
Specific Gravity 1.312
Formula Weight 189.04
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2', 5'-Dichloroacetophenone, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'