missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-4-(trifluoromethyl)pyridine, Thermo Scientific™

Product Code. 10311710
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10311710 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10311710 Supplier Thermo Scientific Maybridge Supplier No. MAY00145DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 89719-92-6
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Molecular Formula C6H2Cl2F3N
MDL Number MFCD09749663
Quantity 1 g
Synonym 2,5-dichloro-4-trifluoromethyl pyridine, pyridine, 2,5-dichloro-4-trifluoromethyl, acmc-20dwro
InChI Key GFTRYJQAASFSIB-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC(Cl)=NC=C1Cl
IUPAC Name 2,5-dichloro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol) 215.98
PubChem CID 26986016
Formula Weight 215.99
Percent Purity 97%
Chemical Name or Material 2, 5-Dichloro-4-(trifluoromethyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.