missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-3-iodopyridine, 98%, Thermo Scientific™

Product Code. 15403787
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942206-23-7
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.882
MDL Number MFCD09037444
InChI Key VYVNSDZZIATDSY-UHFFFAOYSA-N
PubChem CID 42553073
IUPAC Name 2,5-dichloro-3-iodopyridine
SMILES C1=C(C=NC(=C1I)Cl)Cl

Specifications

Melting Point 51°C to 53°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 273.89
Percent Purity 98%
Chemical Name or Material 2,5-Dichloro-3-iodopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'