missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-2,5-dimethylhexane, TRC

Product Code. 30386128
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386128 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386128 Supplier Toronto Research Chemicals Supplier No. TRCD434220250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dichloro-2,5-dimethylhexane
CAS 6223-78-5
Molecular Formula C8H16Cl2
Quantity 250 mg
Synonym 2,5-Dimethyl-2,5-dichlorohexane, NSC 408418
InChI Formula InChI=1S/C8H16Cl2/c1-7(2,9)5-6-8(3,4)10/h5-6H2,1-4H3
SMILES CC(C)(Cl)CCC(C)(C)Cl
IUPAC Name 2,5-dichloro-2,5-dimethylhexane
Molecular Weight (g/mol) 183.12
Formula Weight 182.0629
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.