missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromotoluene, 98%, Thermo Scientific Chemicals

Catalog Number p-102801
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
615-59-8
C7H6Br2
249.93
MFCD00000090
QKEZTJYRBHOKHH-UHFFFAOYSA-N
2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
12006
1,4-dibromo-2-methylbenzene
CC1=C(C=CC(=C1)Br)Br
This item is not returnable. View return policy
615-59-8
C7H6Br2
249.93
MFCD00000090
QKEZTJYRBHOKHH-UHFFFAOYSA-N
2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
12006
1,4-dibromo-2-methylbenzene
CC1=C(C=CC(=C1)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Colorless to Yellow
Density 1.8100g/mL
Boiling Point 135.0°C to 136.0°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3Br2
Refractive Index 1.6010 to 1.6030
Quantity 5 g
Beilstein 05, 308
Specific Gravity 1.81
Solubility Information Solubility in water: immiscible
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 5-Dibromotoluene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dibromotoluene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'