missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromopyridine, TRC

Product Code. 30378743
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378743 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378743 Supplier Toronto Research Chemicals Supplier No. TRCD42547210G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 624-28-2
Molecular Formula C5 H3 Br2 N
Quantity 10 g
Synonym Nicotinic Acid Impurity 13
InChI Formula InChI=1S/C5H3Br2N/c6-4-1-2-5(7)8-3-4/h1-3H
SMILES Brc1ccc(Br)nc1
IUPAC Name 2,5-dibromopyridine
Molecular Weight (g/mol) 236.89
Formula Weight 234.8632
Chemical Name or Material 2,5-Dibromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.