missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromopyridine, 97%

Product Code. 11484343
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484343 50 g 50g
11474343 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484343 Supplier Thermo Scientific Alfa Aesar Supplier No. A10937.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-28-2 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-28-2
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00006221
InChI Key ZHXUWDPHUQHFOV-UHFFFAOYSA-N
Synonym 2,5-dibromo pyridine, pyridine, 2,5-dibromo, 2,5-dibrompyridin, 2,5-dibromo-pyridine, 2,5-dibromopyridin, 3,6-dibromopyridine, 2,5,-dibromopyridine, 2,5-dibrompyridine, 2,5 dibromopyridine, pyridine,5-dibromo
PubChem CID 69353
IUPAC Name 2,5-dibromopyridine
SMILES BrC1=CC=C(Br)N=C1

Specifications

Melting Point 92°C to 96°C
Boiling Point 249°C
Quantity 50 g
Beilstein 109099
Formula Weight 236.9
Percent Purity 97%
Chemical Name or Material 2,5-Dibromopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.