missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzoic acid, 98%

Product Code. 11477377
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477377 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477377 Supplier Thermo Scientific Alfa Aesar Supplier No. A15077.22
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 610-71-9 | C7H4Br2O2 | 279.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 610-71-9
Melting Point 153°C to 157°C
Molecular Formula C7H4Br2O2
MDL Number MFCD00016494
Quantity 100 g
Beilstein 1868193
Synonym benzoic acid, 2,5-dibromo, rarechem fh 1 W 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid
InChI Key SQQKOTVDGCJJKI-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(Br)=CC=C1Br
IUPAC Name 2,5-dibromobenzoic acid
Molecular Weight (g/mol) 279.92
PubChem CID 11891
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.