missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzoic acid, 98%

Product Code. 11457377
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457377 5 g 5g
11467377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457377 Supplier Thermo Scientific Alfa Aesar Supplier No. A15077.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 610-71-9 | C7H4Br2O2 | 279.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-71-9
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016494
InChI Key SQQKOTVDGCJJKI-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
PubChem CID 11891
IUPAC Name 2,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1Br

Specifications

Melting Point 153°C to 157°C
Quantity 5 g
Beilstein 1868193
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.