missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-picoline, 98%, Thermo Scientific™

Product Code. 10094973 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10094973 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10094973 Supplier Thermo Scientific Acros Supplier No. 360950050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3430-18-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 42.0°C to 47.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H5Br2N
MDL Number MFCD02093085
Quantity 5 g
Beilstein 20, V,5, 538
Synonym 2,5-dibromo-3-picoline, 2,5-dibromo-3-methyl-pyridine, pyridine, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methylpyridine, 2,5-dibrom-3-methylpyridin, pubchem1138, acmc-209i6e, ksc495g9h, 2,5-dibromo-3-methyl pyridine
InChI Key LIMXEVCFAUTBCK-UHFFFAOYSA-N
SMILES CC1=CC(Br)=CN=C1Br
IUPAC Name 2,5-dibromo-3-methylpyridine
Molecular Weight (g/mol) 250.92
PubChem CID 817680
Formula Weight 251
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dibromo-3-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dibromo-3-picoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.