missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-n-dodecylthiophene, 97%, Thermo Scientific™

Product Code. 15492358
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492358 1 g 1g
15412368 250 mg 250mg
15402368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492358 Supplier Thermo Scientific Alfa Aesar Supplier No. H55958.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,5-Dibromo-3-n-dodecylthiophene is used as an OLED intermediate, flavor and fragrance intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148256-63-7
Molecular Formula C16H26Br2S
Molecular Weight (g/mol) 410.252
MDL Number MFCD00274315
InChI Key ZIZGNWAMVWNAJT-UHFFFAOYSA-N
PubChem CID 5072754
IUPAC Name 2,5-dibromo-3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=C(SC(=C1)Br)Br

Specifications

Density 1.321
Boiling Point 135°C (2.5 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.53
Quantity 1 g
Synonym thiophene, 2,5-dibromo-3-dodecyl, 2,5-dibromo-3-laurylthiophene, 2,5-dibromo-3-dodecyl-thiophene, 2,5-bromo-3-dodecylthiophene, acmc-209cip, ksc496a6t, bidd:gt0375, thiophene,2,5-dibromo-3-dodecyl, 2,5-bis bromanyl-3-dodecyl-thiophene, 2,5-dibromo-3-dodecylthiophene
Solubility Information Slightly soluble in water.
Formula Weight 410.25
Percent Purity 97%
Chemical Name or Material 2,5-Dibromo-3-n-dodecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.