missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-fluoropyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15467608
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
156772-60-0
C5H2Br2FN
254.884
MFCD06659497
QEENLQKOTDHQEW-UHFFFAOYSA-N
22412832
2,5-dibromo-3-fluoropyridine
C1=C(C(=NC=C1Br)Br)F
This item is not returnable. View return policy
156772-60-0
C5H2Br2FN
254.884
MFCD06659497
QEENLQKOTDHQEW-UHFFFAOYSA-N
22412832
2,5-dibromo-3-fluoropyridine
C1=C(C(=NC=C1Br)Br)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
UN Number UN2811
Synonym 2,5-dibromo-3-fluoro-pyridine, pyridine, 2,5-dibromo-3-fluoro, pubchem17547, acmc-209deo, ksc529c7p, pyridine,2,5-dibromo-3-fluoro, abbypharma ap-30-7594, 2,5-bis bromanyl-3-fluoranyl-pyridine, 2-bromo-3-fluoro-5-bromopyridine
Formula Weight 254.88
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,5-Dibromo-3-fluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'