missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3,4-dinitrothiophene, Thermo Scientific Chemicals

Product Code. 13498498 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13498498 5 g Glass bottle 5g
13488498 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13498498 Supplier Thermo Scientific Acros Supplier No. 278220050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52431-30-8 | C4Br2N2O4S | 331.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52431-30-8
Molecular Formula C4Br2N2O4S
Molecular Weight (g/mol) 331.92
MDL Number MFCD00015537
InChI Key AHGHPBPARMANQD-UHFFFAOYSA-N
Synonym 2,5-dibromo-3,4-dinitro-thiophene, thiophene, 2,5-dibromo-3,4-dinitro, acmc-1apgk, ksc491i5j, 2,5-dibromo-3,4-dinitrothiophene gc
PubChem CID 257024
IUPAC Name 2,5-dibromo-3,4-dinitrothiophene
SMILES [O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O

Specifications

Melting Point 132.0°C to 134.0°C
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 331.94
Chemical Name or Material 2, 5-Dibromo-3, 4-dinitrothiophene
compliance-icons
Product Identifier
  • 2,5-Dibromo-3,4-dinitrothiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.