missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminopyridine dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. p-7044270
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478839 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15478839 Supplier Thermo Scientific Alfa Aesar Supplier No. L05599.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26878-35-3
Molecular Formula C5H9Cl2N3
Molecular Weight (g/mol) 182.048
MDL Number MFCD00040818
InChI Key RWHNUWWUPZKDQP-UHFFFAOYSA-N
Synonym 2,5-diaminopyridine dihydrochloride, pyridine-2,5-diamine dihydrochloride, 2,5-diaminopyridine 2hcl, pyridin-2,5-diamindihydrochlorid, 2,5-diaminopyridinedihydrochloride, pubchem5497, acmc-209gtk, pyridine-2,5-diamine, chloride, chloride
PubChem CID 117889
IUPAC Name pyridine-2,5-diamine;dihydrochloride
SMILES C1=CC(=NC=C1N)N.Cl.Cl

Specifications

Melting Point ∽264°C (decomposition)
Quantity 25 g
Beilstein 3912365
Sensitivity Hygroscopic
Formula Weight 182.05
Percent Purity 97%
Chemical Name or Material 2,5-Diaminopyridine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.