missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminopyridine, 97%

Product Code. 10614565 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10614565 5 g 5g
10282902 1 g 1g
10545914 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10614565 Supplier Thermo Scientific Acros Supplier No. 360690050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4318-76-7 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4318-76-7
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
InChI Key MIROPXUFDXCYLG-UHFFFAOYSA-N
Synonym 2,5-diaminopyridine, 2,5-pyridinediamine, pyridine, 2,5-diamino, diaminopyridine van, unii-h6olz060hi, h6olz060hi, da1, pyridine,5-diamino, 2,5-diamino pyridine, 2,5-diamino-pyridine
PubChem CID 20314
IUPAC Name pyridine-2,5-diamine
SMILES C1=CC(=NC=C1N)N

Specifications

Boiling Point 180.0°C (12.0 mmHg)
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 2, 5-Diaminopyridine
compliance-icons
Product Identifier
  • 2,5-Diaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.