missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminohydroquinone dihydrochloride, 97%

Product Code. p-7026140
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
This item is not returnable. View return policy

Product Code. 11435087

Brand: Thermo Scientific Alfa Aesar B21969.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24171-03-7 | C6H10Cl2N2O2 | 213.058 g/mol

2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

Notes
Air sensitive. It may discolor to black-violet on storage. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24171-03-7
Molecular Formula C6H10Cl2N2O2
Molecular Weight (g/mol) 213.058
MDL Number MFCD00239416
InChI Key NILKAWPWTYPHAH-UHFFFAOYSA-N
Synonym 2,5-diaminobenzene-1,4-diol dihydrochloride, 2,5-diamino-1,4-dihydroxybenzene dihydrochloride, 2,5-diaminohydroquinone dihydrochloride, acmc-20aobi, 1,4-benzenediol, 2,5-diamino-, dihydrochloride, 2,5-diaminobenzene-1,4-diol, chloride, chloride, 1,4-benzenediol,2,5-diamino-,hydrochloride 1:2, 2,5-diaminobenzene-1,4-diol-hydrogen chloride 1/2, 2,5-diaminohydroquinone dihydrochloride, technical at
PubChem CID 16212088
IUPAC Name 2,5-diaminobenzene-1,4-diol;dihydrochloride
SMILES C1=C(C(=CC(=C1O)N)O)N.Cl.Cl

Specifications

Melting Point >300°C
Odor Odorless
Quantity 0.25 g
Sensitivity Air Sensitive
Formula Weight 213.06
Percent Purity 97%
Chemical Name or Material 2,5-Diaminohydroquinone dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.