missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diamino-3-fluoro-4-(trifluoromethyl)pyridine, Thermo Scientific™

Product Code. 10220100
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10220100 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10220100 Supplier Thermo Scientific Maybridge Supplier No. MAY00150DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 675602-90-1
Packaging Amber Glass Bottle
Molecular Formula C6H5F4N3
Quantity 1 g
Synonym 2,5-diamino-3-fluoro-4-trifluoromethyl pyridine, 3-fluoro-4-trifluoromethyl pyridine-2,5-diamine, 2,5-diamino-3-fluoro-4-trifluoromethyl-pyridine
InChI Key SMAJZLPBXNXMJE-UHFFFAOYSA-N
SMILES C1=C(C(=C(C(=N1)N)F)C(F)(F)F)N
IUPAC Name 3-fluoro-4-(trifluoromethyl)pyridine-2,5-diamine
Molecular Weight (g/mol) 195.121
PubChem CID 2758028
Formula Weight 195.12
Percent Purity 95%
Chemical Name or Material 2, 5-diamino-3-fluoro-4-(trifluoromethyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.