missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butylhydroquinone, 99%

Product Code. 10215960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215960 1 kg Plastic bottle 1kg
10404555 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215960 Supplier Thermo Scientific Acros Supplier No. 150840010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-58-4 | C14H22O2 | 222.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-58-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00008825
InChI Key JZODKRWQWUWGCD-UHFFFAOYSA-N
Synonym 2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
PubChem CID 2374
ChEBI CHEBI:41094
IUPAC Name 2,5-ditert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 212°C to 218°C
Color Beige to Beige-Pink
Density 216g/mL
Flash Point 216°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in aqueous alkali,soluble in acetone,alcohol and benzene,practically insoluble in toluene and hexane
Formula Weight 222.33
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Di-tert-butylhydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.