missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butyl-4-methoxyphenol, 97%

Product Code. 15487457
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487457 5 g 5g
15497457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487457 Supplier Thermo Scientific Alfa Aesar Supplier No. H35053.06
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1991-52-2 | C15H24O2 | 236.355 g/mol

2,5-Di-tert-butyl-4-methoxyphenol is used as a non-tumorigenic compound in contrast to DBHQ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Di-tert-butyl-4-methoxyphenol is used as a non-tumorigenic compound in contrast to DBHQ.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1991-52-2
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00274238
InChI Key FLLRQABPKFCXSO-UHFFFAOYSA-N
Synonym 2,5-di-tert-butyl-4-methoxyphenol, 2,5-di-tert-butyl-4-hydroxyanisole, ccris 5220, unii-593e9t2mwh, phenol,2,5-bis 1,1-dimethylethyl-4-methoxy, acmc-1bqnv, spectrum1505039, 2,5-di-t-butyl-4-methoxyphenol, 2,5-di-t-butyl-4-hydroxyanisole, 2,5-di-tert-butyl4-hydroxyanisole
PubChem CID 74812
IUPAC Name 2,5-ditert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=CC(=C(C=C1OC)C(C)(C)C)O

Specifications

Melting Point 99°C to 102°C
Color White to Cream
Quantity 5 g
Formula Weight 236.36
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di-tert-butyl-4-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.