missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Bromo-2-thienyl)pyridine, 97%, Thermo Scientific™

Product Code. 10069081
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10069081 10 g 10g
10232492 1 g 1g
10481114 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10069081 Supplier Thermo Scientific Maybridge Supplier No. CC02210EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 123784-07-6
Molecular Formula C9H6BrNS
Molecular Weight (g/mol) 240.12
MDL Number MFCD00120466
InChI Key AKNKWLQVFAYRRO-UHFFFAOYSA-N
Synonym 2-5-bromo-2-thienyl pyridine, 2-5-bromothiophen-2-yl pyridine, 2-5-bromothien-2-yl pyridine, pyridine,2-5-bromo-2-thienyl, acmc-20mqrz, 5-bromo-2-2-pyridyl thiophene, 2-5-bromo-2-thiophenyl pyridine, 2-bromo-5-2-pyridinyl thiophene, 2-5-bromanylthiophen-2-yl pyridine, 2-5-bromo-thiophen-2-yl-pyridine
PubChem CID 736483
IUPAC Name 2-(5-bromothiophen-2-yl)pyridine
SMILES BrC1=CC=C(S1)C1=CC=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 240.12
Percent Purity 95%
Chemical Name or Material 2-(5-Bromo-2-thienyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.