missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bishydroxymethyl Tetrahydrofuran, TRC

Product Code. 30346962
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346962 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346962 Supplier Toronto Research Chemicals Supplier No. TRCB446375250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Bishydroxymethyl Tetrahydrofuran
CAS 104-80-3
Molecular Formula C6 H12 O3
Quantity 250 mg
Synonym Hexitol, 2,5-anhydro-3,4-dideoxy-, 2,5-Furandimethanol, tetrahydro- (6CI,7CI,8CI), 2,5-Anhydro-3,4-dideoxyhexitol, (Tetrahydrofuran-2,5-diyl)dimethanol, 2,5-Bis(hydroxymethyl)tetrahydrofuran, 2,5-Dihydroxymethyltetrahydrofuran, 2,5-Dihydroxymethyltetrahydrofurane, NSC 40741, Tetrahydro-2,5-furandimethanol
InChI Formula InChI=1S/C6H12O3/c7-3-5-1-2-6(4-8)9-5/h5-8H,1-4H2
SMILES OCC1CCC(CO)O1
IUPAC Name [5-(hydroxymethyl)oxolan-2-yl]methanol
Molecular Weight (g/mol) 132.16
Formula Weight 132.0786
Product Title
Select an issue

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.