missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(tert-butylperoxy)-2,5-dimethyl-3-hexyne (technical grade, stabilized with 40-60% CaCO3), TRC

Product Code. 30480912
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480912 1 g 1g
30467658 5 g 5g
30465688 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480912 Supplier Toronto Research Chemicals Supplier No. TRCL4748951G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-27-5
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.41
Synonym 1,1'-(1,1,4,4-Tetramethyl-2-butyne-1,4-diyl)bis[2-(1,1-dimethylethyl) peroxide], 2,5-Bis(tert-butylperoxy)-2,5-dimethyl-3-hexyne, 2,5-Bis(tert-butylperoxy)-2,5-dimethylhexyne, 2,5-Di(tert-butylperoxy)-2,5-dimethyl-3-hexyne, 2,5-Dimethyl-2,5-bis(tert-butyldioxy)-3-hexyne, 2,5-Dimethyl-2,5-bis(tert-butylperoxy)-3-hexyne, 2,5-Dimethyl-2,5-bis(tert-butylperoxy)hexyne, 2,5-Dimethyl-2,5-di(t-butylperoxy)-3-hexyne, 2,5-Dimethyl-2,5-di(t-butylperoxy)hexyne, 2,5-Dimethyl-2,5-di(t-butylperoxy)hexyne-3, 2,5-Dimethyl-2,5-di(tert-butylperoxy)-3-hexyne, 2,5-Dimethyl-2,5-di(tert-butylperoxy)hexyne-3, 2,5-Dimethyl-2,5-di(tert-butylperoxy)hexyne-3, 2,5-Dimethyl-2,5-di-tert-butylperoxyhexyne, 25B, Aztec DYBP, Cab-O-Cure 2P, D 25, D 25 (peroxide), DYBP, DYBP 85WO, Interox DYBP, Kayahexa YD, Kayahexa YD 50CL 130, L 130 (peroxide), LX 130, Luperco 130XL, Luperox 130, Luperox 130XL45, Luperox XL 130, Lupersol 130, PX 1, PX 1 (peroxide), Perhexyne 2.5B, Perhexyne 2.5B40, Perhexyne 25B, Perhexyne 25B40, Peroxide D 25, Sanperox YPO, Trigonox 101HP, Trigonox 145, Trigonox 145-45B, Trigonox 145B45PD, Trigonox 145E85, Varox 130XL
IUPAC Name 2,5-bis(tert-butylperoxy)-2,5-dimethylhex-3-yne
SMILES CC(C)(C)OOC(C)(C)C#CC(C)(C)OOC(C)(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H30O4/c1-13(2,3)17-19-15(7,8)11-12-16(9,10)20-18-14(4,5)6/h1-10H3
Formula Weight 286.2144
Chemical Name or Material 2,5-Bis(tert-butylperoxy)-2,5-dimethyl-3-hexyne (technical grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.