missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(diethylamino)phenyl-1,3,4-oxadiazole, 98%, Thermo Scientific™

Product Code. 11489936
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489936 100 g 100g
11479936 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489936 Supplier Thermo Scientific Alfa Aesar Supplier No. B20901.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-98-7
Molecular Formula C22H28N4O
Molecular Weight (g/mol) 364.493
MDL Number MFCD00798595
InChI Key UZGVMZRBRRYLIP-UHFFFAOYSA-N
Synonym 2,5-bis 4-diethylaminophenyl-1,3,4-oxadiazole, 2,5-bis 4'-diethylaminophenyl-1,3,4-oxadiazole, benzenamine, 4,4'-1,3,4-oxadiazole-2,5-diyl bis n,n-diethyl, 4-5-4-diethylamino phenyl-1,3,4-oxadiazol-2-yl-n,n-diethylaniline, 4-5-4-diethylamino phenyl 1,3,4-oxadiazol-2-yl phenyl diethylamine, acmc-209dxi, 2,5-bis 4-diethylaminophenyl-1,3,4-oxadiazol, 2,5-bis 4-diethylaminophenyl-1,3,4-oxadizole
PubChem CID 74301
IUPAC Name 4-[5-[4-(diethylamino)phenyl]-1,3,4-oxadiazol-2-yl]-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)N(CC)CC

Specifications

Melting Point 148°C to 151°C
Quantity 100 g
Beilstein 569499
Formula Weight 364.49
Percent Purity 98%
Chemical Name or Material 2,5-Bis(diethylamino)phenyl-1,3,4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.