missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole, 98%, Thermo Scientific™

Catalog Number 15429125
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
146370-52-7
C17H26Cl2O2
333.293
MFCD03093939
TXAVEVGYOGQVAN-UHFFFAOYSA-N
2,5-bis chloromethyl-1-methoxy-4-2-ethylhexyloxy benzene, 1,4-bis chloromethyl-2-2-ethylhexyl oxy-5-methoxybenzene, 2,5-bis chloromethyl-4-2-ethylhexyloxy anisole, acmc-20dwr0, 1,4-bis chloromethyl-2-2-ethylhexoxy-5-methoxy-benzene, 1,4-bis chloromethyl-2-2-ethylhexoxy-5-methoxybenzene, 1,4-bis chloromethyl-2-2-ethylhexyloxy-5-methoxy-benzene, 2,5-bis chloromethyl-1-methoxy-4-2-ethylhexyloxy-benzene
4233009
1,4-bis(chloromethyl)-2-(2-ethylhexoxy)-5-methoxybenzene
CCCCC(CC)COC1=CC(=C(C=C1CCl)OC)CCl
This item is not returnable. View return policy
146370-52-7
C17H26Cl2O2
333.293
MFCD03093939
TXAVEVGYOGQVAN-UHFFFAOYSA-N
2,5-bis chloromethyl-1-methoxy-4-2-ethylhexyloxy benzene, 1,4-bis chloromethyl-2-2-ethylhexyl oxy-5-methoxybenzene, 2,5-bis chloromethyl-4-2-ethylhexyloxy anisole, acmc-20dwr0, 1,4-bis chloromethyl-2-2-ethylhexoxy-5-methoxy-benzene, 1,4-bis chloromethyl-2-2-ethylhexoxy-5-methoxybenzene, 1,4-bis chloromethyl-2-2-ethylhexyloxy-5-methoxy-benzene, 2,5-bis chloromethyl-1-methoxy-4-2-ethylhexyloxy-benzene
4233009
1,4-bis(chloromethyl)-2-(2-ethylhexoxy)-5-methoxybenzene
CCCCC(CC)COC1=CC(=C(C=C1CCl)OC)CCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole is used for conducting polymer precursor. It is also used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN1759
Solubility Information Insoluble in water.
Formula Weight 333.29
Percent Purity 98%
Chemical Name or Material 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'